Assay Details
| Gene Symbol: Prmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866897.3 | XP_004866954.1 | 4 - 5 | 409 | 60 | |
| XM_004866898.2 | XP_004866955.1 | 3 - 4 | 467 | 60 | ||
| XM_013066419.2 | XP_012921873.1 | 4 - 5 | 408 | 60 | ||
More Information
Set Membership: |
|

Prmt1
Genomic Map