Assay Details
| Gene Symbol: LOC101715865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860019.2 | XP_004860076.1 | 2 - 3 | 1230 | 83 | |
| XM_013077898.2 | XP_012933352.1 | 2 - 3 | 1064 | 83 | ||
| XM_013077899.2 | XP_012933353.1 | 2 - 3 | 1035 | 83 | ||
| XM_021240568.1 | XP_021096227.1 | 2 - 3 | 1069 | 83 | ||
| XM_021240570.1 | XP_021096229.1 | 3 - 4 | 1160 | 83 | ||
| XM_021240571.1 | XP_021096230.1 | 2 - 3 | 1069 | 83 | ||
| XM_021240572.1 | XP_021096231.1 | 2 - 3 | 1072 | 83 | ||
More Information
Set Membership: |
|

LOC101715865
Genomic Map