Assay Details
| Gene Symbol: Phkg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856182.2 | XP_004856239.1 | 7 - 8 | 839 | 61 | |
| XM_004856183.2 | XP_004856240.1 | 7 - 8 | 842 | 61 | ||
| XM_021237471.1 | XP_021093130.1 | 7 - 8 | 864 | 61 | ||
More Information
Set Membership: |
|

Phkg2
Genomic Map