Assay Details
| Gene Symbol: Mtss1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842884.3 | XP_004842941.1 | 14 - 15 | 1514 | 66 | |
| XM_004842885.3 | XP_004842942.1 | 13 - 14 | 1409 | 66 | ||
| XM_013071498.2 | XP_012926952.1 | 13 - 14 | 1505 | 66 | ||
| XM_013071499.2 | XP_012926953.1 | 14 - 15 | 1418 | 66 | ||
| XM_013071500.2 | XP_012926954.1 | 8 - 9 | 1188 | 66 | ||
| XM_021256713.1 | XP_021112372.1 | 13 - 14 | 1440 | 66 | ||
| XM_021256714.1 | XP_021112373.1 | 13 - 14 | 1344 | 66 | ||
More Information
Set Membership: |
|

Mtss1l
Genomic Map