Assay Details
Gene Symbol: Nsun7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004849814.3 | XP_004849871.1 | 9 - 10 | 1531 | 98 | |
XM_004849816.3 | XP_004849873.1 | 8 - 9 | 1378 | 98 | ||
XM_021262421.1 | XP_021118080.1 | 9 - 10 | 1531 | 98 | ||
XM_021262422.1 | XP_021118081.1 | 8 - 9 | 1387 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
