Assay Details
| Gene Symbol: LOC101696402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872537.2 | XP_004872594.1 | 4 - 5 | 482 | 75 | |
| XM_004872538.2 | XP_004872595.1 | 3 - 4 | 434 | 75 | ||
| XM_021250332.1 | XP_021105991.1 | 4 - 5 | 482 | 75 | ||
More Information
Set Membership: |
|

LOC101696402
Genomic Map