Assay Details
| Gene Symbol: LOC101696402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872537.2 | XP_004872594.1 | 8 - 9 | 1064 | 74 | |
| XM_004872538.2 | XP_004872595.1 | 7 - 8 | 1016 | 74 | ||
More Information
Set Membership: |
|

LOC101696402
Genomic Map