Assay Details
| Gene Symbol: Slc41a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854160.3 | XP_004854217.1 | 5 - 6 | 771 | 62 | |
| XM_004854161.3 | XP_004854218.1 | 4 - 5 | 671 | 62 | ||
| XM_004854162.3 | XP_004854219.1 | 5 - 6 | 622 | 62 | ||
| XM_004854164.3 | XP_004854221.1 | 4 - 5 | 532 | 62 | ||
| XM_004854165.3 | XP_004854222.1 | 5 - 6 | 702 | 62 | ||
| XM_004854166.3 | XP_004854223.1 | 3 - 4 | 393 | 62 | ||
| XM_004854167.3 | XP_004854224.1 | 5 - 6 | 771 | 62 | ||
| XM_013075647.2 | XP_012931101.1 | 3 - 4 | 878 | 62 | ||
| XM_021266010.1 | XP_021121669.1 | 4 - 5 | 500 | 62 | ||
More Information
Set Membership: |
|

Slc41a3
Genomic Map