Assay Details
| Gene Symbol: Pus3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862579.3 | XP_004862636.1 | 3 - 4 | 828 | 79 | |
| XM_021242802.1 | XP_021098461.1 | 2 - 3 | 472 | 79 | ||
More Information
Set Membership: |
|

Pus3
Genomic Map