Assay Details
| Gene Symbol: Tmlhe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875320.3 | XP_004875377.2 | 2 - 3 | 367 | 64 | |
| XM_013069499.2 | XP_012924953.2 | 2 - 3 | 367 | 64 | ||
More Information
Set Membership: |
|

Tmlhe
Genomic Map