Assay Details
| Gene Symbol: Ahdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850719.3 | XP_004850776.1 | 6 - 7 | 1032 | 127 | |
| XM_021263157.1 | XP_021118816.1 | 5 - 6 | 798 | 127 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004850719.3, XM_021263157.1 transcripts.
Set Membership: |
|

Ahdc1
Genomic Map