Assay Details
| Gene Symbol: Ahdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850719.3 | XP_004850776.1 | 7 - 8 | 5963 | 74 | |
| XM_021263152.1 | XP_021118811.1 | 4 - 5 | 5206 | 74 | ||
| XM_021263154.1 | XP_021118813.1 | 7 - 8 | 6099 | 74 | ||
| XM_021263155.1 | XP_021118814.1 | 7 - 8 | 6102 | 74 | ||
| XM_021263156.1 | XP_021118815.1 | 6 - 7 | 5970 | 74 | ||
| XM_021263157.1 | XP_021118816.1 | 6 - 7 | 5729 | 74 | ||
More Information
Set Membership: |
|

Ahdc1
Genomic Map