Assay Details
| Gene Symbol: Fam83b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868827.2 | XP_004868884.1 | 4 - 5 | 996 | 77 | |
| XM_021247615.1 | XP_021103274.1 | 5 - 6 | 1061 | 77 | ||
| XM_021247616.1 | XP_021103275.1 | 4 - 5 | 1187 | 77 | ||
More Information
Set Membership: |
|

Fam83b
Genomic Map