Assay Details
| Gene Symbol: Jmjd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860770.3 | XP_004860827.1 | 5 - 6 | 1396 | 61 | |
| XM_021241185.1 | XP_021096844.1 | 5 - 6 | 1394 | 61 | ||
More Information
Set Membership: |
|

Jmjd6
Genomic Map