Assay Details
| Gene Symbol: Mdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849985.3 | XP_004850042.1 | 5 - 6 | 697 | 68 | |
| XM_021262551.1 | XP_021118210.1 | 5 - 6 | 602 | 68 | ||
More Information
Set Membership: |
|

Mdh1
Genomic Map