Assay Details
| Gene Symbol: Atp5pb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853716.3 | XP_004853773.1 | 5 - 6 | 680 | 63 | |
| XM_021265662.1 | XP_021121321.1 | 5 - 6 | 676 | 63 | ||
More Information
Set Membership: |
|

Atp5pb
Genomic Map