Assay Details
| Gene Symbol: Rbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845534.3 | XP_004845591.1 | 1 - 2 | 210 | 55 | |
| XM_021258936.1 | XP_021114595.1 | 1 - 2 | 210 | 55 | ||
More Information
Set Membership: |
|

Rbx1
Genomic Map