Assay Details
| Gene Symbol: Rab7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875471.3 | XP_004875528.2 | 425 | 75 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab7a| Gene Symbol: Rab7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875471.3 | XP_004875528.2 | 425 | 75 | ||