Assay Details
| Gene Symbol: Snu13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845507.2 | XP_004845564.1 | 1 - 2 | 165 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004845507.2 transcripts.
Set Membership: |
|

Snu13
Genomic Map