Assay Details
| Gene Symbol: Lrpap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847360.2 | XP_004847417.1 | 5 - 6 | 795 | 79 | |
| XM_021260360.1 | XP_021116019.1 | 5 - 6 | 846 | 79 | ||
More Information
Set Membership: |
|

Lrpap1
Genomic Map