Assay Details
| Gene Symbol: Napsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867200.3 | XP_004867257.3 | 6 - 7 | 952 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Napsa| Gene Symbol: Napsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867200.3 | XP_004867257.3 | 6 - 7 | 952 | 67 | |
Set Membership: |
|