Assay Details
| Gene Symbol: March2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865410.3 | XP_004865467.2 | 4 - 5 | 649 | 82 | |
| XM_004865412.2 | XP_004865469.1 | 4 - 5 | 733 | 82 | ||
| XM_004865413.3 | XP_004865470.1 | 4 - 5 | 1220 | 82 | ||
More Information
Set Membership: |
|

March2
Genomic Map