Assay Details
| Gene Symbol: Mthfd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844754.2 | XP_004844811.2 | 3 - 4 | 502 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mthfd2| Gene Symbol: Mthfd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844754.2 | XP_004844811.2 | 3 - 4 | 502 | 68 | |
Set Membership: |
|