Assay Details
| Gene Symbol: Gtf2h5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013076684.2 | XP_012932138.1 | 2 - 3 | 98 | 104 | |
| XM_021237980.1 | XP_021093639.1 | 2 - 3 | 147 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013076684.2, XM_021237980.1 transcripts.
Set Membership: |
|

Gtf2h5
Genomic Map