Assay Details
| Gene Symbol: Cxcl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848115.3 | XP_004848172.2 | 2 - 3 | 264 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cxcl11| Gene Symbol: Cxcl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848115.3 | XP_004848172.2 | 2 - 3 | 264 | 76 | |
Set Membership: |
|