Assay Details
| Gene Symbol: Kdsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858447.3 | XP_004858504.1 | 5 - 6 | 848 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kdsr| Gene Symbol: Kdsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858447.3 | XP_004858504.1 | 5 - 6 | 848 | 74 | |
Set Membership: |
|