Assay Details
| Gene Symbol: Tnni1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862238.2 | XP_004862295.1 | 6 - 7 | 403 | 67 | |
| XM_004862239.2 | XP_004862296.1 | 6 - 7 | 403 | 67 | ||
| XM_004862241.2 | XP_004862298.1 | 6 - 7 | 403 | 67 | ||
| XM_021242438.1 | XP_021098097.1 | 6 - 7 | 403 | 67 | ||
More Information
Set Membership: |
|

Tnni1
Genomic Map