Assay Details
| Gene Symbol: Atp6v1e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837601.3 | XP_004837658.1 | 6 - 7 | 609 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp6v1e1| Gene Symbol: Atp6v1e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837601.3 | XP_004837658.1 | 6 - 7 | 609 | 74 | |
Set Membership: |
|