Assay Details
| Gene Symbol: S100a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871434.3 | XP_004871491.1 | 2 - 3 | 373 | 64 | |
| XM_004871435.3 | XP_004871492.1 | 2 - 3 | 371 | 64 | ||
More Information
Set Membership: |
|

S100a1
Genomic Map