Assay Details
| Gene Symbol: Tcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843456.3 | XP_004843513.1 | 9 - 10 | 1485 | 64 | |
| XM_021257206.1 | XP_021112865.1 | 9 - 10 | 1479 | 64 | ||
More Information
Set Membership: |
|

Tcn2
Genomic Map