Assay Details
| Gene Symbol: Angptl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865567.2 | XP_004865624.1 | 1 - 2 | 361 | 92 | |
| XM_004865568.2 | XP_004865625.1 | 1 - 2 | 357 | 92 | ||
More Information
Set Membership: |
|

Angptl8
Genomic Map