Assay Details
| Gene Symbol: Eif2s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837368.3 | XP_004837425.1 | 3 - 4 | 521 | 73 | |
| XM_004837369.3 | XP_004837426.2 | 3 - 4 | 442 | 73 | ||
More Information
Set Membership: |
|

Eif2s1
Genomic Map