Assay Details
| Gene Symbol: Eif3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865479.3 | XP_004865536.1 | 7 - 8 | 728 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif3g| Gene Symbol: Eif3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865479.3 | XP_004865536.1 | 7 - 8 | 728 | 55 | |
Set Membership: |
|