Assay Details
| Gene Symbol: Lurap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872954.3 | XP_004873011.1 | 1 - 2 | 882 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lurap1| Gene Symbol: Lurap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872954.3 | XP_004873011.1 | 1 - 2 | 882 | 90 | |
Set Membership: |
|