Assay Details
| Gene Symbol: Hey1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842016.3 | XP_004842073.1 | 2 - 3 | 355 | 56 | |
| XM_004842017.3 | XP_004842074.1 | 2 - 3 | 359 | 56 | ||
More Information
Set Membership: |
|

Hey1
Genomic Map