Assay Details
| Gene Symbol: Mdfic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865271.2 | XP_004865328.1 | 4 - 5 | 784 | 86 | |
| XM_021244769.1 | XP_021100428.1 | 3 - 4 | 515 | 86 | ||
| GenBank mRNA | HQ326667.1 | - | 3 - 4 | 423 | 86 | |
More Information
Set Membership: |
|

Mdfic
Genomic Map