Assay Details
| Gene Symbol: Slc25a21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021246638.1 | XP_021102297.1 | 6 - 7 | 655 | 104 | |
| GenBank mRNA | HQ326683.1 | - | 1011 | 104 | ||
More Information
Set Membership: |
|

Slc25a21
Genomic Map