Assay Details
| Gene Symbol: Atp5mc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013068482.2 | XP_012923936.2 | 2 - 3 | 154 | 89 | |
| XM_021250610.1 | XP_021106269.1 | 2 - 3 | 133 | 89 | ||
| XM_021250611.1 | XP_021106270.1 | 2 - 3 | 99 | 89 | ||
| GenBank mRNA | KM486688.1 | - | 2 - 3 | 94 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013068482.2, XM_021250611.1 transcripts.
Set Membership: |
|

Atp5mc2
Genomic Map