Assay Details
| Gene Symbol: Ube2n | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844968.3 | XP_004845025.1 | 5 | 2212 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2n| Gene Symbol: Ube2n | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844968.3 | XP_004845025.1 | 5 | 2212 | 99 | |
Set Membership: |
|