Assay Details
| Gene Symbol: LOC101715424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863035.3 | XP_004863092.1 | 4 | 1653 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101715424| Gene Symbol: LOC101715424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863035.3 | XP_004863092.1 | 4 | 1653 | 106 | |
Set Membership: |
|