Assay Details
| Gene Symbol: Rab9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870999.2 | XP_004871056.1 | 3 | 508 | 103 | |
| XM_021249113.1 | XP_021104772.1 | 3 | 427 | 103 | ||
More Information
Set Membership: |
|

Rab9a
Genomic Map