Assay Details
| Gene Symbol: LOC101715817 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851993.3 | XP_004852050.1 | 13 - 14 | 2226 | 59 | |
| XM_004851995.3 | XP_004852052.2 | 15 - 16 | 2934 | 59 | ||
More Information
Set Membership: |
|

LOC101715817
Genomic Map