Assay Details
| Gene Symbol: Calu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856392.3 | XP_004856449.1 | 6 - 7 | 959 | 65 | |
| XM_004856393.3 | XP_004856450.1 | 7 - 8 | 1046 | 65 | ||
| XM_013076523.2 | XP_012931977.1 | 6 - 7 | 959 | 65 | ||
| XM_013076524.2 | XP_012931978.2 | 4 - 5 | 617 | 65 | ||
| XM_021237564.1 | XP_021093223.1 | 5 - 6 | 805 | 65 | ||
More Information
Set Membership: |
|

Calu
Genomic Map