Assay Details
| Gene Symbol: Klhl32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862498.2 | XP_004862555.1 | 3 - 4 | 673 | 81 | |
| XM_004862500.2 | XP_004862557.1 | 3 - 4 | 673 | 81 | ||
| XM_013078774.2 | XP_012934228.1 | 3 - 4 | 673 | 81 | ||
| XM_021242673.1 | XP_021098332.1 | 3 - 4 | 673 | 81 | ||
| XM_021242675.1 | XP_021098334.1 | 3 - 4 | 673 | 81 | ||
| XM_021242676.1 | XP_021098335.1 | 3 - 4 | 673 | 81 | ||
More Information
Set Membership: |
|

Klhl32
Genomic Map