Assay Details
| Gene Symbol: Map7d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866167.3 | XP_004866224.1 | 14 - 15 | 1994 | 70 | |
| XM_004866168.3 | XP_004866225.1 | 13 - 14 | 1897 | 70 | ||
| XM_004866169.3 | XP_004866226.1 | 13 - 14 | 1871 | 70 | ||
| XM_013066046.2 | XP_012921500.1 | 14 - 15 | 1964 | 70 | ||
| XM_013066047.2 | XP_012921501.1 | 12 - 13 | 1774 | 70 | ||
| XM_021245317.1 | XP_021100976.1 | 10 - 11 | 1672 | 70 | ||
More Information
Set Membership: |
|

Map7d2
Genomic Map