Assay Details
| Gene Symbol: Adgrg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021242552.1 | XP_021098211.1 | 22 - 23 | 9109 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adgrg4| Gene Symbol: Adgrg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021242552.1 | XP_021098211.1 | 22 - 23 | 9109 | 91 | |
Set Membership: |
|