Assay Details
| Gene Symbol: Mcur1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021260624.1 | XP_021116283.1 | 2 - 3 | 592 | 72 | |
| XM_021260626.1 | XP_021116285.1 | 2 - 3 | 604 | 72 | ||
| XM_021260627.1 | XP_021116286.1 | 2 - 3 | 587 | 72 | ||
| XM_021260628.1 | XP_021116287.1 | 2 - 3 | 584 | 72 | ||
| XM_021260629.1 | XP_021116288.1 | 2 - 3 | 585 | 72 | ||
| XM_021260630.1 | XP_021116289.1 | 2 - 3 | 597 | 72 | ||
| XM_021260631.1 | XP_021116290.1 | 2 - 3 | 594 | 72 | ||
| XM_021260632.1 | XP_021116291.1 | 2 - 3 | 591 | 72 | ||
| XM_021260633.1 | XP_021116292.1 | 2 - 3 | 591 | 72 | ||
| XM_021260634.1 | XP_021116293.1 | 2 - 3 | 606 | 72 | ||
More Information
Set Membership: |
|

Mcur1
Genomic Map