Assay Details
| Gene Symbol: Tyrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838291.2 | XP_004838348.1 | 4 - 5 | 1049 | 83 | |
| XM_004838292.2 | XP_004838349.1 | 4 - 5 | 1049 | 83 | ||
More Information
Set Membership: |
|

Tyrp1
Genomic Map