Assay Details
| Gene Symbol: Nol4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840574.1 | XP_004840631.1 | 9 - 10 | 1621 | 59 | |
| XM_004840575.3 | XP_004840632.1 | 6 - 7 | 1021 | 59 | ||
| XM_021254886.1 | XP_021110545.1 | 9 - 10 | 1382 | 59 | ||
More Information
Set Membership: |
|

Nol4l
Genomic Map