Assay Details
| Gene Symbol: Slc16a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857355.3 | XP_004857412.2 | 1 - 2 | 1086 | 94 | |
| XM_021238208.1 | XP_021093867.1 | 2 - 3 | 829 | 94 | ||
| XM_021238209.1 | XP_021093868.1 | 2 - 3 | 990 | 94 | ||
| XM_021238210.1 | XP_021093869.1 | 2 - 3 | 843 | 94 | ||
| XM_021238211.1 | XP_021093870.1 | 3 - 4 | 885 | 94 | ||
| XM_021238212.1 | XP_021093871.1 | 2 - 3 | 824 | 94 | ||
More Information
Set Membership: |
|

Slc16a11
Genomic Map